Learn More
Solvent Red 26, Dr. Ehrenstorfer
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Solvent Red 26 |
| CAS | 4477-79-6 |
| Analyte or Component Names | Solvent Red 26 |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H22 N4 O |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 2-Naphthalenol, 1-[[2,5-dimethyl-4-[(2-methylphenyl)azo]phenyl]azo]- (9CI), C.I. Solvent Red 26 (7CI,8CI), 1-[2-[2,5-Dimethyl-4-[2-(2-methylphenyl)diazenyl]phenyl]diazenyl]-2-naphthalenol, 1-[4-(o-Tolylazo)-2,5-xylylazo]-2-naphthol, Akrodye Oil Red 1, C.I. 26120, Calco Oil Red A 1700, Calco Oil Red N 1700, Calco Oil Red Z 1700, Calcogas Rocket Red, NSC 115877, Oil Red 2144, Oil Red EGN, Oil Red GPD, Oil Red OB, Oil Red PEL, Solvent Red 26 |
| InChI Formula | InChI=1S/C25H22N4O/c1-16-8-4-7-11-21(16)26-27-22-14-18(3)23(15-17(22)2)28-29-25-20-10-6-5-9-19(20)12-13-24(25)30/h4-15,30H,1-3H3 |
| Solution Type | Neat |
| SMILES | Cc1ccccc1N=Nc2cc(C)c(cc2C)N=Nc3c(O)ccc4ccccc34 |
| Show More |