missing translation for 'onlineSavingsMsg'
Learn More

Abcam Sodium Oxamate, Lactate Dehydrogenase Inhibitor, Solid, 1 g

Product Code. 32120211
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32120211 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32120211 Supplier Abcam Supplier No. ab1456431g
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity sodium oxamate, a weak lactate dehydrogenase inhibitor that suppresses glycolysis and gluconeogenesis.

  • Molecular weight: 111.03 Da
  • Purity: >99%
  • Form: Solid
  • Inhibitor: Weak lactate dehydrogenase inhibitor (IC50 values are 600 and 4400 μM in H460 and A549 cells respectively)
  • Key biological effects: Inhibits glycolysis and gluconeogenesis, decreases extracellular acidification rate
  • Analog: Pyruvate (ab145036) analog
  • CAS Number: 565-73-1
  • Composition: 100% Sodium oxamate

Specifications

Buffer 100% sodium oxamate
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Target Molecule Lactate Dehydrogenase
Formulation 100% Sodium Oxamate
Quantity 1 g
For Use With (Application) Inhibition of lactate dehydrogenase, glycolysis and gluconeogenesis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.