Learn More
Sodium oxalate, For ACS analysis, 99.96%, MP Biomedicals™
Product Code. 11424652 Shop All MP Biomedical Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424652 500 g 500g
11414652 100 g 100g
11434652 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11424652 Supplier MP Biomedicals Supplier No. 0219143990
Chemical Identifiers
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O
Specifications
Color White
Density 2.34 (water = 1)
Quantity 500 g
Formula Weight 134
Physical Form Crystalline Powder
Chemical Name or Material Sodium oxalate, For ACS analysis