Learn More
Sodium hexafluoroaluminate, 99.9% (metals basis)
Description
Sodium aluminum hexafluoride is used as a solvent (or flux) for electrolysis aluminum oxides such as bauxite, whitener for enamels and an opacifier for glass and in the industrial production of aluminum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1000°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 99.9% (metals basis) |
| Solubility Information | Soluble in water(0.4g/L). |
| Formula Weight | 209.95 |
| Odor | Odorless |
| Density | 2.9 |
| Chemical Name or Material | Sodium hexafluoroaluminate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.