accessibility menu, dialog, popup

UserName

CAS RN 13775-53-6

FFFFFFNaNaNaAl

CAS RN 13775-53-6

IUPAC Name: aluminium(3+) trisodium hexafluoride
Synonyms: sodium hexafluoroaluminate
Molecular Weight (g/mol): 209.94
Molecular Formula: AlF6Na3
InChi Key: REHXRBDMVPYGJX-UHFFFAOYSA-H
SMILES: [F-].[F-].[F-].[F-].[F-].[F-].[Na+].[Na+].[Na+].[Al+3]
Physical Form 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (5)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)