Learn More
Sodium Cumenesulfonate (~93% purity), TRC
Product Code. 30424220
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424220 10 g -
30418467 25 g 25g
30416156 50 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30424220 Supplier Toronto Research Chemicals Supplier No. TRCS65507010G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28348-53-0
Molecular Formula C9H11O3SNa
Molecular Weight (g/mol) 222.24
Synonym Sodium Cumolsulfonate, Eltesol SC 40, Sodium Isopropylbenzenesulfonate, Stepanate SCS, Taycatox N 5040, Lutensit TC-CS 40
SMILES [Na+].CC(C)c1ccc(cc1)S(=O)(=O)[O-]
Specifications
Quantity 10 g
Chemical Name or Material Sodium Cumenesulfonate