CAS RN 28348-53-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Sodium Cumenesulfonate (~93% purity), TRC
CAS: 28348-53-0 Molecular Formula: C9H11O3SNa Molecular Weight (g/mol): 222.24 Synonym: Sodium Cumolsulfonate,Eltesol SC 40,Sodium Isopropylbenzenesulfonate,Stepanate SCS,Taycatox N 5040,Lutensit TC-CS 40 SMILES: [Na+].CC(C)c1ccc(cc1)S(=O)(=O)[O-]