missing translation for 'onlineSavingsMsg'
Learn More

Sodium Calcium Exchanger 1/NCX1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

280.00€ - 624.00€

Specifications

Antigen Sodium Calcium Exchanger 1/NCX1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18299363
View Documents
Novus Biologicals
NBP2-56483
100 μL
624.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18688588
View Documents
Novus Biologicals
NBP2-56483-25ul
25 μL
280.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium Calcium Exchanger 1/NCX1 Polyclonal specifically detects Sodium Calcium Exchanger 1/NCX1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Sodium Calcium Exchanger 1/NCX1
Polyclonal
Rabbit
Human
FLJ37694, FLJ43417, Na(+)/Ca(2+)-exchange protein 1, sodium/calcium exchanger 1, solute carrier family 8 (sodium/calcium exchanger), member 1
SLC8A1
IgG
Affinity Purified
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
6546
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RHAADQARKAVSMHEVNTEVTENDPVSKIFFEQGTYQCLE
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.