Learn More
Gravis Single Use Crimping Bottles
Product Code. p-8009932
Click to view available options
Capacity (Metric):
10 mL
15 mL
24 mL
50 mL
100 mL
Color:
Amber
Clear
Unit Size:
Pack of 130
Pack of 168
Pack of 252
Pack of 88
Product Code. 11793813
Brand: Gravis F018124585
Only null left
Add to basket
Edge
Add to basket
Provides excellent storage for liquids in the laboratory. Gravis Single Use Crimping Bottles are excellent for holding serums.
  • Available in clear or amber glass for light-sensitive materials
  • Manufactured from soda lime glass
  • Neck diameter of 20mm
Specifications
Capacity (Metric) 10 mL
Color Clear
Diameter (Metric) Outer Neck 20 mm
Material Soda Lime Glass
No. per Pack 168Pack
Quantity 168Pack
Product Type Crimp Bottle