missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD2/Mn-SOD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB100-1969-25ug
This item is not returnable.
View return policy
Description
SOD2/Mn-SOD Polyclonal specifically detects SOD2/Mn-SOD in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Gerbil, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| SOD2/Mn-SOD | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% Glycerol with 0.09% Sodium Azide | |
| EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects approx 25 kDa. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:5000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
| P04179 | |
| SOD2 | |
| Recombinant Human Mn SOD Protein | |
| 25 μg | |
| Cancer, Diabetes Research, Tumor Suppressors | |
| 6648 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction