missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98875-50ul
This item is not returnable.
View return policy
Description
SOCS-4 Polyclonal antibody specifically detects SOCS-4 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SOCS-4 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:1000-1:4000 | |
| CIS4, DKFZp686J1568, SOCS-4, SOCS7, SOCS-7, SSI4, STAT4, suppressor of cytokine signaling 4, Suppressor of cytokine signaling 7SH2 domain containing SOCS box protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SOCS-4. | |
| 50 μg | |
| Cell Biology, Signal Transduction | |
| 122809 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction