missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX15 Mouse, anti-Human, CoraLite Plus 488, Clone: 1A10B7, Proteintech
Shop All Proteintech ProductsDescription
The SNX15 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SNX15. This antibody recognizes human antigen. The SNX15 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
| Antigen | SNX15 |
| Applications | Immunofluorescence |
| Clone | 1A10B7 |
| Concentration | 1000 μg/mL |
| Conjugate | CoraLite Plus 488 |
| Gene Symbols | HSAF001435, SNX15, sorting nexin 15 |
| Host Species | Mouse |
| Quantity | 20 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 29907 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?