missing translation for 'onlineSavingsMsg'
Learn More

SNURPORTIN1 Antibody (SNUPN/7363R), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18560369 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18560369 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18560369 Supplier Novus Biologicals Supplier No. NBP320884AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SNURPORTIN1 Monoclonal antibody specifically detects SNURPORTIN1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNURPORTIN1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SNUPN/7363R
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias KPNBL, RNA U transporter 1, RNA, U transporter 1, RNUT1, snurportin 1, Snurportin1, SNURPORTIN-1, SPN1
Host Species Rabbit
Immunogen Recombinant fragment (around aa 900-1100) of human SNURPORTIN1 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10073
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.