missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
483.00€
Specifications
| Antigen | SNRPF |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
SNRPF Polyclonal specifically detects SNRPF in Human samples. It is validated for Western Blot.Specifications
| SNRPF | |
| Polyclonal | |
| Rabbit | |
| PBSCF, Sm protein F, small nuclear ribonucleoprotein polypeptide F, SMF, Sm-Fsmall nuclear ribonucleoprotein F, snRNP-F | |
| SNRPF | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 6636 | |
| Synthetic peptides corresponding to SNRPF(small nuclear ribonucleoprotein polypeptide F) The peptide sequence was selected from the middle region of SNRPF. Peptide sequence GYMNMQLANTEEYIDGALSGHLGEVLIRCNNVLYIRGVEEEEEDGEMRE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title