missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNRPD2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97664-100ul
This item is not returnable.
View return policy
Description
SNRPD2 Polyclonal antibody specifically detects SNRPD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SNRPD2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:500 | |
| small nuclear ribonucleoprotein D2 polypeptide (16.5kD), small nuclear ribonucleoprotein D2 polypeptide 16.5kDa, small nuclear ribonucleoprotein Sm D2, Sm-D2SMD2, snRNP core protein D2, SNRPD1 | |
| Produced in rabbits immunized with E. coli-derived Human SNRPD2 fragment. | |
| 100 μg | |
| Stem Cell Markers | |
| 6633 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction