missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SnoN Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77306-0.025mg
This item is not returnable.
View return policy
Description
SnoN Polyclonal specifically detects SnoN in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| SnoN | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| SKI-like oncogene, Ski-related oncogene, ski-related oncogene snoN, Ski-related protein, SnoA, SnoI, SnoN, SNOski-like protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6498 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| NP_005405 | |
| SKIL | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human SnoN. The immunogen is located within the first 50 amino acids of SnoN. Amino Acid Squence: KKLNGMGDDGSPPAKK | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction