missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNF8 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67696-1-IG-20UL
This item is not returnable.
View return policy
Description
The SNF8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SNF8. This antibody recognizes human antigen. The SNF8 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| SNF8 | |
| Monoclonal | |
| 500 μg/mL | |
| Dot3; EAP30; VPS22 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 29 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2C10F2 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 11267 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction