missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SNAP23 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
Description
An un-tagged recombinant protein corresponding to the amino acids 1-211 of Human SNAP23 The Recombinant Human SNAP23 Protein is derived from E. coli. The Recombinant Human SNAP23 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_003816.2 |
| Concentration | 1 mg/mL |
| For Use With (Application) | ELISA |
| Formulation | 20mM Tris pH 8.0 |
| Gene ID (Entrez) | 8773 |
| Purification Method | Protein |
| Quantity | 0.05 mg |
| Source | Recombinant SNAP23 protein expressed in E. coli |
| Immunogen | SNAP23, 1-211aa, Human, Recombinant, E. coli (NP_003816.2). |
| Storage Requirements | Store at -80°C. Avoid freeze-thaw cycles. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction