missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Snail Antibody (CL3700) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61631MFV500
This item is not returnable.
View return policy
Description
Snail Monoclonal antibody specifically detects Snail in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Snail | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| dJ710H13.1, Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAH, SNAI1, SNAIL, snail 1 homolog, snail 1 zinc finger protein, snail family zinc finger 1, snail homolog 1, SNAIL1, zinc finger protein SNAI1 | |
| This antibody was developed using a recombinant protein derived from O95863, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Developmental Biology, Hypoxia, Lipid and Metabolism, Metastasis, Transcription Factors and Regulators | |
| 6615 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL3700 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction