missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMPD4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00€ - 506.00€
Specifications
| Antigen | SMPD4 |
|---|---|
| Dilution | Western Blot 1:200-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605240
|
Novus Biologicals
NBP2-93253-0.02ml |
0.02 mL |
216.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681451
|
Novus Biologicals
NBP2-93253-0.1ml |
0.1 mL |
506.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMPD4 Polyclonal antibody specifically detects SMPD4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SMPD4 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 55627 | |
| IgG | |
| Affinity purified |
| Western Blot 1:200-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| KIAA1418, NET13, Neutral Sphingomyelinase 3, Neutral Sphingomyelinase III, NSMASE3, NSMASE-3, SKNY Protein, Sphingomyelin Phosphodiesterase 4, Sphingomyelin Phosphodiesterase 4, Neutral Membrane (Neutral Sphingomyelinase-3) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 600-800 of human SMPD4 (NP_060421.2). ITQAKHTAKSISDQCAESPAGHSFLSWLGFSSMDTNGSYTANDLDEMGQDSVRKTDEYLEKALEYLRQIFRLSEAQLRQFTLALGTTQDENGKKQLPDCIVGEDGLILTPLGRYQIINGLRRFEIEYQGDPELQPIRSYEIASLVRTLFRLSSAINHRFAGQMAALCSRDDFLGSFCRYHLTEPGLASRHLLSPVGRRQVA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title