missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Smoothened Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

213.00€ - 508.00€

Specifications

Antigen Smoothened
Dilution Western Blot : 1:400
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30066809
View Documents
Novus Biologicals™
NBP3-30919-100ug
100 μg
508.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30067238
View Documents
Novus Biologicals™
NBP3-30919-25ug
25 μg
213.00€
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Smoothened Polyclonal antibody specifically detects Smoothened in Human samples. It is validated for Western Blot
Specifications

Specifications

Smoothened
Western Blot
Unconjugated
Rabbit
GPCR, Growth and Development, Neuronal Cell Markers
Phosphate buffered saline
6608
IgG
Antigen Affinity-purified
Western Blot : 1:400
Polyclonal
Purified
RUO
Human
Gx, Protein Gx, SMOHseven transmembrane helix receptor, smoothened (Drosophila) homolog, smoothened homolog, smoothened homolog (Drosophila)
Synthetic peptide derived from the human Smoothened protein
Primary
Store at -20°C. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.