Learn More
Thermo Scientific™ SMART Digest™ Trypsin Kits
Product Code. 15668693 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
Bulk Low pH Magnetic Resin
Bulk Low pH Resin
Bulk Magnetic Resin
Bulk Non-magnetic Resin
Resin in Pre-filled PCR Tubes
Soluble Trypsin
Type:
Soluble Trypsin Kit
Soluble Trypsin Kit - 10 Pk.
Soluble Trypsin Kit - Low pH
Trypsin Kit
Includes:
Low pH Resin, Collection Plate
Resin, Buffer
Resin, Buffer, Collection Plate, Filter Plate
Resin, Buffer, Collection Plate, SOLAμ SPE Plate
Resin, Collection Plate
Resin, Collection Plate, Filter Plate
Resin, Collection Plate, SOLAμ SPE Plate
Soluble Trypsin, Buffer, Collection Plate
Soluble Trypsin, Low pH Buffer, Collection Plate
Unit Size:
1 set
10 sets
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Type Includes Unit Size
15668693 Soluble Trypsin Soluble Trypsin Kit Soluble Trypsin, Buffer, Collection Plate Each
15382360 Resin in Pre-filled PCR Tubes Trypsin Kit Resin, Collection Plate 1 set
16434839 Bulk Low pH Resin Trypsin Kit Resin, Collection Plate 1 set
15994417 Resin in Pre-filled PCR Tubes Trypsin Kit Low pH Resin, Collection Plate Each
15317298 Resin in Pre-filled PCR Tubes Trypsin Kit Resin, Collection Plate, SOLAμ SPE Plate Each
16444839 Bulk Low pH Magnetic Resin Trypsin Kit Resin, Collection Plate 1 set
15357350 Resin in Pre-filled PCR Tubes Trypsin Kit Resin, Collection Plate, Filter Plate Each
30130374 Soluble Trypsin Soluble Trypsin Kit - Low pH Soluble Trypsin, Low pH Buffer, Collection Plate 1 set
15791377 Bulk Magnetic Resin Trypsin Kit Resin, Buffer 1 set
15648663 Bulk Magnetic Resin Trypsin Kit Resin, Buffer Each
15658663 Bulk Non-magnetic Resin Trypsin Kit Resin, Buffer, Collection Plate, Filter Plate Each
15668663 Bulk Magnetic Resin Trypsin Kit Resin, Buffer, Collection Plate, Filter Plate Each
15678663 Bulk Non-magnetic Resin Trypsin Kit Resin, Buffer, Collection Plate, SOLAμ SPE Plate Each
15688663 Bulk Magnetic Resin Trypsin Kit Resin, Buffer, Collection Plate, SOLAμ SPE Plate Each
30090382 Soluble Trypsin Soluble Trypsin Kit - 10 Pk. Soluble Trypsin, Buffer, Collection Plate 10 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 15668693 Supplier Thermo Scientific™ Supplier No. 60113101
 Restricted Item
Estimated Shipment: 22-12-2026
Obtain fast, simple, and highly reproducible trypsin protein digestion for peptide characterization and quantitation in biopharmaceutical protein research.
Obtain high quality analytical results from protein digests using Thermo Scientific™ SMART Digest™ Kits. A significant advance in sample preparation for biopharmaceutical protein research, the kits provide fast and simple protein digestion with high reproducibility, high sensitivity, and high levels of data quality in a format that's compatible with automation. The SMART Digest Kits are a significant improvement over current in-solution protein digestion technologies, which are not reproducible, have poor sensitivity, and employ protracted methodologies not amenable to automation.
Trypsin Protein Digestion
SMART Digest kits enable you to characterize and quantitate proteins faster, easier, and quicker than ever before. This version of the kit uses trypsin, which cleaves peptides on the C-terminal side of lysine and arginine amino acid residues.
Easy and simple
  • Significantly increased speed of sample processing
  • Simple process
  • Easily automate entire protocol for high-throughput processing
Reliable Results
  • Increased reproducibility over existing protocols
  • Higher sensitivity
  • Higher data confidence
Significantly Reduces Sample Preparation Time
Typically it takes less than 60 minutes (dependent on sample complexity) to achieve full digestion, whereas an in-solution digest can take up to 24 hours.
Cleaner Samples
Reduction in the number of chemicals used and the immobilized trypsin design helps mitigate against chemically induced post translational modifications (PTMs) and autolysis. This results in results which are easier to interpret.
Couple with SOLAμ SPE Plates for Desalting
When used with the SMART Digest kits for desalting, Thermo Scientific™ SOLAμ™ SPE solid phase extraction plates provide a fast, easy-to-use, robust, and easily automatable sample preparation solution for analysis of biomolecules.
TRUSTED_SUSTAINABILITY
Specifications
Type Soluble Trypsin Kit
Description SMART Digest Trypsin Kit, Soluble
Format Soluble Trypsin
Enzyme Trypsin
Cleavage C-terminal side of lysine and arginine amino acids
Capture None
Product Line SMART Digest
For Use With (Application) Peptide Mapping
Includes Soluble Trypsin, Buffer, Collection Plate