missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00€ - 470.00€
Specifications
| Antigen | SMAD6 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615290
|
Novus Biologicals
NBP2-93787-0.02ml |
0.02 mL |
188.00€
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643221
|
Novus Biologicals
NBP2-93787-0.1ml |
0.1 mL |
470.00€
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SMAD6 Polyclonal antibody specifically detects SMAD6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| SMAD6 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Signaling Pathway | |
| PBS (pH 7.3), 50% glycerol | |
| 4091 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| hSMAD6, HsT17432, MAD homolog 6, MADH6SMAD, mothers against DPP homolog 6 (Drosophila), MADH7, mothers against decapentaplegic homolog 6, Mothers against decapentaplegic, drosophila, homolog of, 6, Mothers against DPP homolog 6, SMAD 6, SMAD family member 6MAD, mothers against decapentaplegic homolog 6 (Drosophila), SMAD, mothers against DPP homolog 6, Smad6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 257-496 of human Smad6 (NP_005576.3). PTVCCNPYHFSRLCGPESPPPPYSRLSPRDEYKPLDLSDSTLSYTETEATNSLITAPGEFSDASMSPDATKPSHWCSVAYWEHRTRVGRLYAVYDQAVSIFYDLPQGSGFCLGQLNLEQRSESVRRTRSKIGFGILLSKEPDGVWAYNRGEHPIFVNSPTLDAPGGRALVVRKVPPGYSIKVFDFERSGLQHAPEPDAADGPYDPNSVRISFAKGWGPCYSRQFITSCPCWLEILLNNPR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title