missing translation for 'onlineSavingsMsg'
Learn More

SLITRK1 Antibody (1), FITC , Novus Biologicals™

Product Code. 18632851 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18632851 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18632851 Supplier Novus Biologicals Supplier No. NBP289435F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

SLITRK1 Monoclonal antibody specifically detects SLITRK1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLITRK1
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate FITC
Formulation PBS
Gene Alias FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SLITRK1 (NP_443142.1; Met 1-Ser 616).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 114798
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.