Learn More
Thermo Scientific™ SLIPFREE™ HPLC Column Double Connectors
Product Code. 10771844 Shop All Thermo Scientific Products
Click to view available options
Type:
Double Flexible Connector
Diameter (English) Inner:
0.005 in.
Length (Metric):
15 cm
Unit Size:
Each
Product Code. 10771844
Brand: Thermo Scientific™ 30215FLEX
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ensure leak-free, void-free column connections and optimize LC performance with self-adjusting Thermo Scientific™ SLIPFREE™ HPLC Column Double Connectors.
Ensure excellent column connections and optimal HPLC performance with Thermo Scientific™ SLIPFREE™ HPLC Column Double Connectors. Compatible with all column end-fittings, these universal, self-adjusting connectors provide leak- and void-free column connections. Fingertight connections up to 10000psi ensure ease of use. They are ideal for supercritical fluid columns (SFC). SLIPFREE connectors are available in a variety of configurations and designs for a broad range of applications.
Leak-Free, Void-Free Connections:
  • Pushes tubing and ferrule into end-fitting
  • Self adjusts to fit each new column connection
  • Fingertight connection up to 10000 psi
Flexible:
  • Fits all brands of columns
  • Available in single and double configurations
  • Available in a range of internal diameters to suit multiple applications
  • PEEK-collared and long-neck designs available for specific applications
TRUSTED_SUSTAINABILITY
Specifications
Type Double Flexible Connector
Diameter (English) Inner 0.005 in.
Description Double Flexible; 0.005 in. ID x 15 cm
For Use With (Equipment) Microbore/Smallbore Column
Length (Metric) 15 cm