missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC7A5/LAT1 Antibody (EP3-1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20115-0.2mg
This item is not returnable.
View return policy
Description
SLC7A5/LAT1 Monoclonal antibody specifically detects SLC7A5/LAT1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Functional Assay
Specifications
| SLC7A5/LAT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunofluorescence, Immunoprecipitation, Functional Assay | |
| EP3-1 | |
| Unconjugated | |
| 4F2 LC, CD98, CD98LC, D16S469EL-type amino acid transporter 1, E16, Integral membrane protein E16, large neutral amino acids transporter 1, large neutral amino acids transporter small subunit 1, LAT1hLAT1, MPE16CD98 light chain, sodium-independent neutral amino acid transporter LAT1,4F2LC, solute carrier family 7 (cationic amino acid transporter, y+ system), member 5,4F2 light chain, Solute carrier family 7 member 5, y+ system cationic amino acid transporter | |
| Immunised mice and constructed IgG1 Fab phage display libraries from the spleen cells. SLC7A5/LAT1-reactive recombinant Fab fragments were retrieved by epitope peptide panning. | |
| 0.2 mg | |
| Cancer | |
| 8140 | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction