missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A4/5-HTTLPR/Serotonin transporter Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05546F
This item is not returnable.
View return policy
Description
SLC6A4/5-HTTLPR/Serotonin transporter Polyclonal antibody specifically detects SLC6A4/5-HTTLPR/Serotonin transporter in Human, Mouse, Rat, Bovine samples. It is validated for Immunofluorescence
Specifications
| SLC6A4/5-HTTLPR/Serotonin transporter | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| 5HT transporter, 5-HTT, hSERT, HTT5-hydroxytryptamine transporter, Na+/Cl- dependent serotonin transporter, OCD1, SERT, sodium-dependent serotonin transporter, solute carrier family 6 (neurotransmitter transporter, serotonin), member 4,5-HTTLPR, Solute carrier family 6 member 4,5HTT | |
| KSITPETPTEIPCGDIRLNAV coupled to KLH | |
| 0.1 mL | |
| Neuroscience | |
| 6532 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunofluorescence | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction