missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A2/NET/Noradrenaline transporter Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 624.00€
Specifications
| Antigen | SLC6A2/NET/Noradrenaline transporter |
|---|---|
| Dilution | Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622428
|
Novus Biologicals
NBP2-62704-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18635206
|
Novus Biologicals
NBP2-62704 |
100 μg |
624.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC6A2/NET/Noradrenaline transporter Polyclonal antibody specifically detects SLC6A2/NET/Noradrenaline transporter in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SLC6A2/NET/Noradrenaline transporter | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Neuroscience | |
| PBS (pH 7.2) and 40% Glycerol | |
| 6530 | |
| IgG | |
| Protein A purified |
| Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| NAT1neurotransmitter transporter, NET, NET1sodium-dependent noradrenaline transporter, Norepinephrine transporter, SLC6A5, solute carrier family 6 (neurotransmitter transporter, noradrenalin), member 2, Solute carrier family 6 member 2, solute carrier family 6 member 5 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MLLARMNPQVQPENNGADTGPEQPLRARKTAELLVVKERNGVQCLLA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title