missing translation for 'onlineSavingsMsg'
Learn More

SLC39A4/ZIP4 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

201.00€ - 482.00€

Specifications

Antigen SLC39A4/ZIP4
Dilution Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18638652
View Documents
Novus Biologicals
NBP2-94856-0.02ml
0.02 mL
201.00€
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18618122
View Documents
Novus Biologicals
NBP2-94856-0.1ml
0.1 mL
482.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC39A4/ZIP4 Polyclonal antibody specifically detects SLC39A4/ZIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

SLC39A4/ZIP4
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Endocrinology, Signal Transduction
PBS (pH 7.3), 50% glycerol
55630
IgG
Affinity purified
Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin
Polyclonal
Purified
RUO
Human, Mouse, Rat
acrodermatitis enteropathica, zinc-deficiency type, FLJ20327, MGC74741, solute carrier family 39 (zinc transporter), member 4, Solute carrier family 39 member 4, zinc transporter ZIP4, ZIP-4, ZIP4AEZ, Zrt- and Irt-like protein 4
Recombinant fusion protein containing a sequence corresponding to amino acids 23-327 of human SLC39A4 (NP_570901.2). SPPAGLLSLLTSGQGALDQEALGGLLNTLADRVHCANGPCGKCLSVEDALGLGEPEGSGLPPGPVLEARYVARLSAAAVLYLSNPEGTCEDARAGLWASHADHLLALLESPKALTPGLSWLLQRMQARAAGQTPKTACVDIPQLLEEAVGAGAPGSAGGVLAALLDHVRSGSCFHALPSPQYFVDFVFQQHSSEVPMTLAELSALMQRLGVGREAHSDHSHRHRGASSRDPVPLISSSNSSSVWDTVCLSARDVMAAYGLSEQAGVTPEAWAQLSPALLQQQLSGACTSQSRPPVQDQLSQSERY
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.