missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC39A3 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
564.00€
Specifications
| Antigen | SLC39A3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | PerCP |
| Form | Purified |
Description
SLC39A3 Polyclonal antibody specifically detects SLC39A3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SLC39A3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| solute carrier family 39 (zinc transporter), member 3, zinc transporter ZIP3, ZIP-3, ZIP3Solute carrier family 39 member 3, Zrt- and Irt-like protein 3 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of human ZIP3. The immunogen is located within amino acids 110 - 160 of ZIP3. Amino Acid Squence: NAGSDVGSDSEYESPF | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot | |
| PerCP | |
| Rabbit | |
| Cancer, Cell Cycle and Replication | |
| PBS | |
| 29985 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title