missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35D2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38040
This item is not returnable.
View return policy
Description
SLC35D2 Polyclonal specifically detects SLC35D2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SLC35D2 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q76EJ3 | |
| SLC35D2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: YSFLTLSSQLKPKPVGEENICLD | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| HFRC, HFRC1UDP-N-acetylglucosamine transporter, Homolog of Fringe connection protein 1, MGC117215, MGC142139, Solute carrier family 35 member D2, solute carrier family 35, member D2, SQV7Lfringe connection, SQV7-like protein, UDP-galactose transporter-related protein 8, UDP-N-acetylglucosamine/UDP-glucose/GDP-mannose transporter, UGTREL8, UGTrel8hfrc | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 11046 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction