missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC34A2 Recombinant Human Monoclonal Antibody (7B6)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560823
This item is not returnable.
View return policy
Description
SLC34A2 Recombinant Monoclonal Antibody for ELISA
SLC34A2 encodes a protein that is a pH-sensitive sodium-dependent phosphate transporter. Phosphate uptake is increased at lower pH. Defects in this gene are a cause of pulmonary alveolar microlithiasis. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| SLC34A2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O95436 | |
| SLC34A2 | |
| Recombinant Human SLC34A2 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 7B6 | |
| Unconjugated | |
| SLC34A2 | |
| AA536683; D5Ertd227e; Na(+)/Pi cotransporter 2B; Na(+)-dependent phosphate cotransporter 2B; NAPI IIb; NaPi-2b; NaPi3b; NAPI-3B; NAPI-IIb; Npt2b; NPTIIb; rNaPi IIb; Slc34a2; Sodium/phosphate cotransporter 2B; Sodium-dependent phosphate transport protein 2B; sodium-phosphate transport protein 2B; solute carrier family 34 (sodium phosphate), member 2; solute carrier family 34 (type II sodium/phosphate contransporter), member 2; solute carrier family 34 (type II sodium/phosphate cotransporter), member 2; solute carrier family 34 member 2; type II sodium-dependent phosphate transporter 3b; type IIb Na/Picotransporter; type IIb sodium-phosphate transporter | |
| Human | |
| Affinity chromatography | |
| RUO | |
| 10568 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction