Learn More
SLC31A1/CTR1 Antibody (S06-7G0), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19918-50ul
 View more versions of this product
Product Code. 18675484
  • 421.00€ / 50µL
Description
SLC31A1/CTR1 Monoclonal antibody specifically detects SLC31A1/CTR1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SLC31A1/CTR1
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S06-7G0
Unconjugated
Copper transporter 1, COPT1high affinity copper uptake protein 1, CTR1MGC75487, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1
A synthetic peptide of human SLC31A1/CTR1 (Uniprot # O15431)
50 μg
Neuroscience, Neurotransmission, Protein Kinase
1317
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.