missing translation for 'onlineSavingsMsg'
Learn More

SLC26A7 Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

129.00€ - 349.00€

Specifications

Antigen SLC26A7
Concentration 250 μg/mL
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30236247
View Documents
Proteintech
17654-1-AP-150UL
150 μL
ONLINE EXCLUSIVE PRICE
410.00€
349.00€ / 150µL
Save 61.00€
14% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237986
View Documents
Proteintech
17654-1-AP-20UL
20 μL
ONLINE EXCLUSIVE PRICE
145.00€
129.00€ / 20µL
Save 16.00€
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SLC26A7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC26A7. This antibody recognizes human, mouse, rat antigen. The SLC26A7 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications

Specifications

SLC26A7
Western Blot, ELISA
Unconjugated
Rabbit
Human, Mouse, Rat
MGC126268; SUT2
IgG
Antigen affinity purification
Polyclonal Antibody
250 μg/mL
Polyclonal
Liquid
RUO
115111
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.