missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A42 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67819-1-IG-150UL
This item is not returnable.
View return policy
Description
The SLC25A42 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC25A42. This antibody recognizes human antigen. The SLC25A42 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| SLC25A42 | |
| Monoclonal | |
| 1000 μg/mL | |
| MGC26694 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human | |
| 33-35 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 2F7G10 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 284439 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction