missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A36 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | SLC25A36 |
|---|---|
| Clone | 1E5E4 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237269
|
Proteintech
67896-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30238069
|
Proteintech
67896-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SLC25A36 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC25A36. This antibody recognizes human, mouse, rat antigen. The SLC25A36 antibody has been validated for the following applications: WB, IHC, IF/ICC, ELISA analysis.Specifications
| SLC25A36 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 55186 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1E5E4 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| DKFZp564C053; FLJ10618 | |
| IgG1 | |
| Protein G purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title