missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC22A2/OCT2 Antibody (CL0628) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52941PE
This item is not returnable.
View return policy
Description
SLC22A2/OCT2 Monoclonal antibody specifically detects SLC22A2/OCT2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| SLC22A2/OCT2 | |
| Monoclonal | |
| PE | |
| hOCT2, MGC32628, OCT2Organic cation transporter 2, solute carrier family 22 (organic cation transporter), member 2, solute carrier family 22 member 2 | |
| This antibody was developed using a recombinant protein derived from O15244, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL0628 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 6582 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction