Learn More
Invitrogen™ SLC19A1 Monoclonal Antibody (890513)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561955
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Transport of folate compounds into mammalian cells can occur via receptor-mediated or carrier-mediated mechanisms. A functional coordination between these 2 mechanisms has been proposed to be the method of folate uptake in certain cell types. Methotrexate (MTX) is an antifolate chemotherapeutic agent that is actively transported by the carrier-mediated uptake system. RFC1 plays a role in maintaining intracellular concentrations of folate.
Specifications
| SLC19A1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| P41440 | |
| SLC19A1 | |
| NS0 mouse myeloma cell line transfected with human Reduced Folate Carrier/SLC19A1, Met1-Gln591. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 890513 | |
| Unconjugated | |
| SLC19A1 | |
| AI323572; CHMD; FLOT1; folate transporter 1; FOLT; IFC1; IFC-1; Intestinal folate carrier 1; intestinal folate carrier protein; methotrexate carrier 1; methotrexate carrier 2; methotrexate carrier 5; methotrexate carrier 6; MTX1; MTX-1; Placental folate transporter; reduced folate carrier 1; Reduced folate carrier protein; Reduced folate transporter; Reduced folate transporter 1; REFC; RFC; RFC1; RFC-1; RFT-1; SLC19A1; solute carrier family 19 (folate transporter), member 1; solute carrier family 19 (sodium/hydrogen exchanger), member 1; solute carrier family 19 member 1; solute carrier family 19, member 1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6573 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |