missing translation for 'onlineSavingsMsg'
Learn More

SLC14A1 Antibody (888418) [Biotin], Novus Biologicals™

Product Code. 18614245 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18614245 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18614245 Supplier Novus Biologicals Supplier No. FAB8238B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SLC14A1 Monoclonal antibody specifically detects SLC14A1 in Human samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC14A1
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 888418
Conjugate Biotin
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias FLJ33745, HsT1341, HUT11, JKFLJ41687, RACH1, solute carrier family 14 (urea transporter), member 1 (Kidd blood group), Solute carrier family 14 member 1, truncated urea transporter, urea transporter 1, urea transporter JK glycoprotein, Urea transporter, erythrocyte, urea transporter-B1, UT1UT-B1, UTEblood group Kidd urea transporter
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human SLC14A1, Accession # Q13336
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6563
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.