missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
SKIP Polyclonal antibody specifically detects SKIP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | SKIP |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | EC 3.1.3, EC 3.1.3.56, inositol polyphosphate-5-phosphatase K, PPS, skeletal muscle and kidney enriched inositol phosphatase, Skeletal muscle and kidney-enriched inositol phosphatase, SKIPinositol polyphosphate 5-phosphatase K |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 149-448 of human INPP5K (NP_057616.2). HLPPHISNNYQRLEHFDRILEMQNCEGRDIPNILDHDLIIWFGDMNFRIEDFGLHFVRESIKNRCYGGLWEKDQLSIAKKHDPLLREFQEGRLLFPPTYKFDRNSNDYDTSEKKRKPAWTDRILWRLKRQPCAGPDTPIPPASHFSLSLRGYSSHMTYGISDHKPVSGTFDLELKPLVSAPLIVLMPEDLWTVENDMMVSYSSTSDFPSSPWDWIGLYKVGLRDVNDYVSYAWVGDSKVSCSDNLNQVYIDISNIPTTEDEFLLCYYSNSLRSVVGISRPFQIPPGSLREDPLGEAQPQI |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?