missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIX2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94801-0.02ml
This item is not returnable.
View return policy
Description
SIX2 Polyclonal antibody specifically detects SIX2 in Human, Rat samples. It is validated for Western Blot
Specifications
| SIX2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| homeobox protein SIX2, sine oculis homeobox (Drosophila) homolog 2, Sine oculis homeobox homolog 2, sine oculis homeobox homolog 2 (Drosophila), SIX homeobox 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-174 of human SIX2 (NP_058628.3). SIWDGEETSYCFKEKSRSVLREWYAHNPYPSPREKRELAEATGLTTTQVSNWFKN | |
| 0.02 mL | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10736 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction