missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sirtuin 1/SIRT1 Polyclonal specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockout Validated, Knockdown Validated.
Specifications
Specifications
| Antigen | Sirtuin 1/SIRT1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:150, Knockout Validated, Knockdown Validated |
| Formulation | PBS, 0.1% BSA, and 50% Glycerol with 0.05% Sodium Azide |
| Gene Accession No. | Q96EB6, Q923E4 |
| Gene Alias | EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 |
| Gene Symbols | SIRT1 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?