missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIRPB1 Monoclonal Antibody (308908)
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA561098
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
Reconstitute at 0.5 mg/mL in sterile PBS.
SIRP beta 1 is a type I transmembrane protein belonging to the SIRP family within the Ig superfamily. Members of this family are characterized by an extracellular region containing a V-set Ig domain with a J-like sequence and two C1-set Ig domains. Unlike SIRP alpha that has cytoplasmic ITIM domains, SIRP beta 1 possesses positively charged residues that allow association with ITAM motif-containing adaptor molecules. SIRP beta 1 is expressed on cells of monocyte, macrophage or dendritic lineages.
Specifica
| SIRPB1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O00241 | |
| SIRPB1 | |
| Mouse myeloma cell line NS0-derived recombinant human SIRPB1/CD172b, Gly26-Ala369 (Arg53His and Ala363Pro). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry | |
| 308908 | |
| Unconjugated | |
| SIRPB1 | |
| 9930027N05Rik; CD172 antigen-like family member B; CD172b; CD172b antigen; DKFZp686A05192; signal regulatory protein beta 1; signal-regulatory protein beta 1; signal-regulatory protein beta 1A; signal-regulatory protein beta-1; Signal-regulatory protein beta-1 isoform 3; SIRP BETA 1; SIRP beta 1 cell surface protein; Sirpb; SIRPB1; Sirpb1a; SIRP-beta; SIRP-beta B; SIRP-BETA-1; SIRP-beta-1 isoform 3 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto