missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRP gamma/CD172g Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99880-50ul
This item is not returnable.
View return policy
Description
SIRP gamma/CD172g Polyclonal antibody specifically detects SIRP gamma/CD172g in Human samples. It is validated for Western Blot, ELISA
Specifications
| SIRP gamma/CD172g | |
| Polyclonal | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-B2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPgamma, SIRP-gamma | |
| Produced in rabbits immunized with purified, recombinant Human SIRP gamma/CD172g (Accession#: NP_061026.2; Met 1-Ser 364) | |
| 50 μg | |
| Cell Cycle and Replication | |
| 55423 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction