Learn More
Thermo Scientific™ SilTite™ Ferrule Kits
Product Code. 10792897 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific DSQ GC-MS Interface
Thermo Scientific ISQ GC-MS Interface
Thermo Scientific TRACE 1300/1310 GC SSL Injectors
Diameter (Metric) Inside:
0.1 to 0.25 mm
0.32 mm
0.53 mm
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Diameter (Metric) Inside Unit Size
10792897 Thermo Scientific TRACE 1300/1310 GC SSL Injectors 0.1 to 0.25 mm Pack of 10
10177904 Thermo Scientific ISQ GC-MS Interface 0.1 to 0.25 mm Pack of 10
10578405 Thermo Scientific ISQ GC-MS Interface 0.32 mm Pack of 10
10609965 Thermo Scientific ISQ GC-MS Interface 0.53 mm Pack of 10
10661196 Thermo Scientific TRACE 1300/1310 GC SSL Injectors 0.32 mm Pack of 10
10086154 Thermo Scientific TRACE 1300/1310 GC SSL Injectors 0.53 mm Pack of 10
10007084 Thermo Scientific DSQ GC-MS Interface 0.1 to 0.25 mm Pack of 10
10097294 Thermo Scientific DSQ GC-MS Interface 0.32 mm Pack of 10
10724556 Thermo Scientific DSQ GC-MS Interface 0.53 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10792897 Supplier Thermo Scientific™ Supplier No. 290MA215
Big Savings for Better Separation
Use promo code 27992
View all promotions
Estimated Shipment: 29-10-2026
Add to basket
Add to basket
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits.
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits. The ferrule forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule Kit
Description 290 MA215
For Use With (Equipment) Thermo Scientific TRACE 1300/1310 GC SSL Injectors
Material SilTite Metal
Includes N/A
Diameter (Metric) Inside 0.1 to 0.25 mm