Learn More
Macherey-Nagel™ Silicone Septa
Click to view available options
Diameter (Metric) Outer:
9 mm
12 mm
13 mm
Unit Size:
Pack of 50
Specifications
Specifications
| Type | Septum |
| Diameter (Metric) Outer | 13 mm |
| Hardness | 45° Shore A |
| Temperature (Metric) Max | 250°C |
| Color | Clear |
| Material | Silicone |
| Quantity | 50 Pack |