Learn More
Macherey-Nagel™ Silica N/UV254 Adsorbents
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
Specifications
Specifications
| Quantity | 1 kg |
| Product Type | Silica N/UV254 Adsorbent |
| Particle Size | 2 to 20 μm |
| Material | Silica |
| Pore Volume | 0.75 mL/g |
| Pore Size | 60 Å |