Learn More
Macherey-Nagel™ SILCEL-Mix-25 cellulose and silica mixed layer on glass plates
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Pack of 25
Description
Separate preservatives and other antimicrobial compounds. SILCEL-Mix-25 Cellulose and Silica Mixed Layer on Glass Plates is the perfect choice in TLC separations.
- Mixed layer of cellulose and silica
- Recommended for separation of preservatives
- Recommended for separation of antimicrobial compounds
Specifications
Specifications
| Material | Cellulose and Silica |
| Quantity | 25 Pk. |
| Type | Glass Plate |
| Fluorescent Indicator | UV254 |
| Thickness (Metric) | 0.25 mm |
| Dimensions (L x W) | 20 x 20 cm |