missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sildenafil Citrate, Mikromol™
Description
Sildenafil Citrate
Specifications
Specifications
| Chemical Name or Material | Sildenafil Citrate |
| CAS | 171599-83-0 |
| Analyte or Component Names | Sildenafil Citrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H30 N6 O4 S . C6 H8 O7 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Sildenafil Citrate, 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Piperazine, 1-[[3-(4,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methyl-, 2-hydroxy-1,2,3-propanetricarboxylate (1:1) (9CI), 1-[[3-(6,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methylpiperazine, 2-hydroxy-1,2,3-propanetricarboxylate (1:1), Actra-Rx, Alisiga, Apodefil, Caverta 100, Edegra, Kamagra, Maxifort, Niagra Actra-Rx, Penegra, Sildegra, Sildenafil citrate, Sildigra, Siligra, Sollevare, Tonafil, UK 92480, UK 92480-10, Vega, Vega (pharmaceutical), Viagra, Vigrex, Zenegra |
| InChI Formula | InChI=1S/C22H30N6O4S.C6H8O7/c1-5-7-17-19-20(27(4)25-17)22(29)24-21(23-19)16-14-15(8-9-18(16)32-6-2)33(30,31)28-12-10-26(3)11-13-28;7-3(8)1-6(13,5(11)12)2-4(9)10/h8-9,14H,5-7,10-13H2,1-4H3,(H,23,24,29);13H,1-2H2,(H,7,8)(H,9,10)(H,11,12) |
| SMILES | CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(C)CC4.OC(=O)CC(O)(CC(=O)O)C(=O)O |
| IUPAC Name | 5-[2-ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one;2-hydroxypropane-1,2,3-tricarboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?