Learn More
Brand™ Silberbrand™ Eterna™ AR-GLAS™ Class B Measuring Pipets
Product Code. 12075302 Shop All Brand Products
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
25 mL
Graduations:
0.01 mL
0.02 mL
0.05 mL
0.1 mL
Tolerance:
±0.008 mL
±0.010 mL
±0.015 mL
±0.05 mL
±0.08 mL
±0.15 mL
Color:
2 x Green
Black
Blue
Green
Orange
Red
White
Yellow
Unit Size:
Pack of 12
Pack of 6
Product Code. 12075302
Brand: Brand™ 27071
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Manufactured from AR-GLAS™ soda lime glass, this Class B Measuring Pipet is ideal in most research, scientific, and other laboratory environments.
  • Manufactured from AR-GLAS™ Soda lime glass.
  • Type 3, with zero value at the top.
  • Amber stain marking.
  • Class B.
  • Compliant with DIN EN ISO 835 standards.
  • Calibrated to deliver (TD, Ex).
Specifications
Graduations 0.1 mL
Tolerance ±0.010 mL
Color Red
Volume (Metric) 1 mL